Learn More
Biomatik Corporation Mouse Complement C1q tumor necrosis factor-related protein 3 (C1QTNF3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36633
Encompass_Preferred
Mouse Complement C1Q Tumor Necrosis Factor-Related Protein 3 (C1Qtnf3) Elisa Kit for detecting C1Q And Tumor Necrosis Factor Related Protein 3 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9ES30; C1qtnf3. Target Synonyms: C1qtnf3; Cors26; Ctrp3; Complement C1q tumor necrosis factor-related protein 3; Collagenous repeat-containing sequence 26 kDa protein; CORS26; Secretory protein CORS26. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8451