Learn More
Biomatik Corporation Mouse Complement fragment 3a, C3a ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36653
Encompass_Preferred
Mouse Complement Fragment 3A, C3A Elisa Kit for detecting Complement Fragment 3A, C3A in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8454