Learn More
Biomatik Corporation Mouse Complement fragment 5a, C5a ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36656
Encompass_Preferred
Mouse Complement Fragment 5A, C5A Elisa Kit for detecting Complement Fragment 5A, C5A in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 39-2500pg/ml. Sensitivity: 9.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8456