Learn More
Biomatik Corporation Mouse Corticotropin-releasing Factor (CRF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36664
Encompass_Preferred
Mouse Corticotropin-Releasing Factor (Crf) Elisa Kit for detecting Corticotropin Releasing Hormone in serum, plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.5-40pg/ml. Sensitivity: 0.675pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q8CIT0; Crh. Target Synonyms: Crh; Gm1347Corticoliberin; Corticotropin-releasing factor; CRF; Corticotropin-releasing hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8460