Learn More
Biomatik Corporation Mouse Cramp ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36669
Encompass_Preferred
Mouse Cramp Elisa Kit for detecting Cathelicidin Antimicrobial Peptide in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P51437; Camp. Target Synonyms: Camp; Cnlp; CrampCathelicidin antimicrobial peptide; Cathelin-like protein; CLP) [Cleaved into: Cathelin-related antimicrobial peptide; Cramp)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8462