Learn More
Biomatik Corporation Mouse Creatine Kinase MB isoenzyme, CK-MB ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36673
Encompass_Preferred
Mouse Creatine Kinase Mb Isoenzyme, Ck-Mb Elisa Kit for detecting Creatine Kinase Mb Isoenzyme, Ck-Mb in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.25-80mU/ml. Sensitivity: 0.312mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8464