Learn More
Biomatik Corporation Mouse Creatine kinase M-type (CKM) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36675
Encompass_Preferred
Mouse Creatine Kinase M-Type (Ckm) Elisa Kit for detecting Creatine Kinase, Muscle in serum, plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100mU/ml. Sensitivity: 0.39mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P07310; Ckm. Target Synonyms: Ckm; Ckmm; Creatine kinase M-type; EC 2.7.3.2; Creatine kinase M chain; Creatine phosphokinase M-type; CPK-M; M-CK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8466