Learn More
Biomatik Corporation Mouse cross linked N-telopeptide of type collagen, NTX ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36677
Encompass_Preferred
Mouse Cross Linked N-Telopeptide Of Type I Collagen, Ntx Elisa Kit for detecting Cross Linked N-Telopeptide Of Type I Collagen, Ntx in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200nM BCE. Sensitivity: 0.78nM BCE. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Target Synonyms: cross linked N-telopeptide of type 1 collagen; NTX; cross linked N-telopeptide of type I collagen; NTX. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8467