Learn More
Biomatik Corporation Mouse C-X-C motif chemokine 3 (CXCL3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36689
Encompass_Preferred
Mouse C-X-C Motif Chemokine 3 (Cxcl3) Elisa Kit for detecting Chemokine (C-X-C Motif) Ligand 3 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q6W5C0; Cxcl3. Target Synonyms: Cxcl3; Dcip1; Gm1960C-X-C motif chemokine 3; Dendritic cell inflammatory protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8470