Learn More
Biomatik Corporation Mouse Desmosine ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36733
Encompass_Preferred
Mouse Desmosine Elisa Kit for detecting Desmosine in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8477