Learn More
Biomatik Corporation Mouse diamine oxidase, DAO ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36734
Encompass_Preferred
Mouse Diamine Oxidase, Dao Elisa Kit for detecting Amiloride Binding Protein 1 (Amine Oxidase (Copper-Containing)) in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000mU/ml. Sensitivity: 3.9mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8JZQ5; Aoc1. Target Synonyms: Aoc1; Abp1Amiloride-sensitive amine oxidase [copper-containing]; DAO; Diamine oxidase; EC 1.4.3.22; Amiloride-binding protein 1; Amine oxidase copper domain-containing protein 1; Histaminase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8478