Learn More
Biomatik Corporation Mouse Dihydrotestosterone, DHT ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36741
Encompass_Preferred
Mouse Dihydrotestosterone, Dht Elisa Kit for detecting Dihydrotestosterone, Dht in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.1-20nmol/L. Sensitivity: 0.05nmol/l. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8479