Learn More
Biomatik Corporation Mouse dopamine, DA ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36768
Encompass_Preferred
Mouse Dopamine, Da Elisa Kit for detecting Dopamine, Da in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 5-1000pg/ml. Sensitivity: 2.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8480