Learn More
Biomatik Corporation Mouse Ectodysplasin-A (EDA) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36777
Encompass_Preferred
Mouse Ectodysplasin-A (Eda) Elisa Kit for detecting Ectodysplasin A in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O54693; Eda. Target Synonyms: Eda; Ed1; Ta; Ectodysplasin-A; EDA protein homolog; Tabby protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8482