Learn More
Biomatik Corporation Mouse Endosialin (CD248) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36791
Encompass_Preferred
Mouse Endosialin (Cd248) Elisa Kit for detecting Cd248 Molecule, Endosialin in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.81-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q91V98; Cd248. Target Synonyms: Cd248; Tem1; Endosialin; Tumor endothelial marker 1; CD antigen CD248. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8485