Learn More
Biomatik Corporation Mouse Endothelial nitric oxide synthase, eNOS ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36796
Encompass_Preferred
Mouse Endothelial Nitric Oxide Synthase, Enos Elisa Kit for detecting Nitric Oxide Synthase 3 (Endothelial Cell) in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20IU/ml. Sensitivity: 0.078IU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P70313; Nos3. Target Synonyms: Nos3; EcnosNitric oxide synthase; endothelial; EC 1.14.13.39; Constitutive NOS; cNOS; EC-NOS; Endothelial NOS; eNOS; NOS type III; NOSIII. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8487