Learn More
Biomatik Corporation Mouse Eotaxin 2 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36803
Encompass_Preferred
Mouse Eotaxin 2 Elisa Kit for detecting Chemokine (C-C Motif) Ligand 24 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9JKC0; Ccl24. Target Synonyms: Ccl24; Scya24; C-C motif chemokine 24; Eosinophil chemotactic protein 2; Eotaxin-2; Small-inducible cytokine A24. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8489