Learn More
Biomatik Corporation Mouse Ephrin-A1 (EFNA1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36814
Encompass_Preferred
Mouse Ephrin-A1 (Efna1) Elisa Kit for detecting Ephrin-A1 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 93.75-6000pg/ml. Sensitivity: 23.43pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P52793; Efna1. Target Synonyms: Efna1; Epgl1; Epl1; Lerk1; Ephrin-A1; EPH-related receptor tyrosine kinase ligand 1; LERK-1; Immediate early response protein B61) [Cleaved into: Ephrin-A1; secreted form]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8492