Learn More
Biomatik Corporation Mouse Epidermal growth factor-like protein 6 (EGFL6) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36817
Encompass_Preferred
Mouse Epidermal Growth Factor-Like Protein 6 (Egfl6) Elisa Kit for detecting Egf-Like-Domain, Multiple 6 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 93.75-6000pg/ml. Sensitivity: 23.43pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9JJZ5; Egfl6. Target Synonyms: Egfl6; Maeg; Epidermal growth factor-like protein 6; EGF-like protein 6; MAM and EGF domains-containing gene protein; Protein W80. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8493