Learn More
Biomatik Corporation Mouse Fibroblast growth factor 15 (FGF15) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36861
Encompass_Preferred
Mouse Fibroblast Growth Factor 15 (Fgf15) Elisa Kit for detecting Fibroblast Growth Factor 15(Fgf15) in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 3.9pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: O35622; Fgf15. Target Synonyms: FGF-15; Fgf15; FGF15_MOUSE; FGF19; Fibroblast growth factor 15. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8501