Learn More
Biomatik Corporation Mouse Filaggrin (FLG) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36889
Encompass_Preferred
Mouse Filaggrin (Flg) Elisa Kit for detecting Filaggrin in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P11088; Flg. Target Synonyms: FlgFilaggrin; Fragment. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8502