Learn More
Biomatik Corporation mouse follicle-stimulating hormone, FSH ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36894
Encompass_Preferred
Mouse Follicle-Stimulating Hormone, Fsh Elisa Kit for detecting Follicle-Stimulating Hormone, Fsh in serum, plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 4-140mIU/ml. Sensitivity: 2.5mIU/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: Q60687; Fshb. Target Synonyms: FshbFollitropin subunit beta; Follicle-stimulating hormone beta subunit; FSH-B; FSH-beta; Follitropin beta chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8503