Learn More
Biomatik Corporation Mouse galanin prepropeptide (GAL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36912
Encompass_Preferred
Mouse Galanin Prepropeptide (Gal) Elisa Kit for detecting Galanin Prepropeptide in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P47212; Gal. Target Synonyms: Gal; Galn; Galanin peptides [Cleaved into: Galanin; Galanin message-associated peptide; GMAP)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8508