Learn More
Biomatik Corporation Mouse Gastrin ELISA Kit, 96 tests
Catalog No. 501498510
Encompass_Preferred
Mouse Gastrin Elisa Kit for detecting Gastrin in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.25-80pg/ml. Sensitivity: 0.31pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P48757; Gast. Target Synonyms: Gast; Gas; Gastrin [Cleaved into: Gastrin-71; G71); Big gastrin; Gastrin-34; G34); Gastrin]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8510 Supplier Biomatik Corporation Supplier No. EKC36932
Mouse Gastrin Elisa Kit for detecting Gastrin in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.25-80pg/ml. Sensitivity: 0.31pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P48757; Gast. Target Synonyms: Gast; Gas; Gastrin [Cleaved into: Gastrin-71; G71); Big gastrin; Gastrin-34; G34); Gastrin]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.