Learn More
Biomatik Corporation Mouse ghrelin (GHRL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36938
Encompass_Preferred
Mouse Ghrelin (Ghrl) Elisa Kit for detecting Ghrelin (Ghrl) in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.37-1000pg/ml. Sensitivity: 1.37pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9EQX0; Ghrl. Target Synonyms: Ghrl; MtlrpAppetite-regulating hormone; Growth hormone secretagogue; Growth hormone-releasing peptide; Motilin-related peptide; Protein M46) [Cleaved into: Ghrelin; Obestatin]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8513