Learn More
Biomatik Corporation Mouse Glucose-6-phosphate isomerase (GPI) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36952
Encompass_Preferred
Mouse Glucose-6-Phosphate Isomerase (Gpi) Elisa Kit for detecting Glucose Phosphate Isomerase in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.195ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P06745; Gpi. Target Synonyms: Gpi; Gpi1Glucose-6-phosphate isomerase; GPI; EC 5.3.1.9; Autocrine motility factor; AMF; Neuroleukin; NLK; Phosphoglucose isomerase; PGI; Phosphohexose isomerase; PHI. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8516