Learn More
Biomatik Corporation Mouse Glutamate decarboxylase 1 (GAD1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC36959
Encompass_Preferred
Mouse Glutamate Decarboxylase 1 (Gad1) Elisa Kit for detecting Glutamate Decarboxylase 1 (Brain, 67Kda) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.25-400pg/ml. Sensitivity: 1.56pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P48318; Gad1. Target Synonyms: Gad1; Gad67Glutamate decarboxylase 1; EC 4.1.1.15; 67 kDa glutamic acid decarboxylase; GAD-67; Glutamate decarboxylase 67 kDa isoform. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8519