Learn More
Biomatik Corporation Mouse Glypican-3 (GPC-3) ELISA Kit, 96 tests
Catalog No. 501498523
Encompass_Preferred
Mouse Glypican-3 (Gpc-3) Elisa Kit for detecting Glypican 3 in serum, plasma, cell culture supernates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8CFZ4; Gpc3. Target Synonyms: Gpc3; Glypican-3 [Cleaved into: Glypican-3 alpha subunit; Glypican-3 beta subunit]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8523 Supplier Biomatik Corporation Supplier No. EKC36984
Mouse Glypican-3 (Gpc-3) Elisa Kit for detecting Glypican 3 in serum, plasma, cell culture supernates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q8CFZ4; Gpc3. Target Synonyms: Gpc3; Glypican-3 [Cleaved into: Glypican-3 alpha subunit; Glypican-3 beta subunit]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.