Learn More
Biomatik Corporation Mouse gonadotropin-releasing hormone, GnRH ELISA Kit, 96 tests
Catalog No. 501498524
Encompass_Preferred
Mouse Gonadotropin-Releasing Hormone, Gnrh Elisa Kit for detecting Gonadotropin-Releasing Hormone 1 (Luteinizing-Releasing Hormone) in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P13562; Gnrh1. Target Synonyms: Gnrh1; GnrhProgonadoliberin-1; Progonadoliberin I) [Cleaved into: Gonadoliberin-1; Gonadoliberin I; Gonadotropin-releasing hormone I; GnRH-I; Luliberin I; Luteinizing hormone-releasing hormone I; LH-RH I); Prolactin release-inhibiting factor 1; Prolactin release-inhibiting factor I)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8524 Supplier Biomatik Corporation Supplier No. EKC36986
Mouse Gonadotropin-Releasing Hormone, Gnrh Elisa Kit for detecting Gonadotropin-Releasing Hormone 1 (Luteinizing-Releasing Hormone) in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P13562; Gnrh1. Target Synonyms: Gnrh1; GnrhProgonadoliberin-1; Progonadoliberin I) [Cleaved into: Gonadoliberin-1; Gonadoliberin I; Gonadotropin-releasing hormone I; GnRH-I; Luliberin I; Luteinizing hormone-releasing hormone I; LH-RH I); Prolactin release-inhibiting factor 1; Prolactin release-inhibiting factor I)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.