Learn More
Biomatik Corporation Mouse growth hormone, GH ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37002
Encompass_Preferred
Mouse Growth Hormone, Gh Elisa Kit for detecting Growth Hormone 1 in serum, plasma, cell culture supernates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200pg/ml. Sensitivity: 0.78pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P06880; Gh1. Target Synonyms: Gh1; Gh; Somatotropin; Growth hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8527