Learn More
Biomatik Corporation Mouse Heat Shock Protein 90, Hsp-90 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37025
Encompass_Preferred
Mouse Heat Shock Protein 90, Hsp-90 Elisa Kit for detecting Heat Shock Protein 90Kda Alpha (Cytosolic), Class A Member 1 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P07901; Hsp90aa1. Target Synonyms: Hsp90aa1; Hsp86; Hsp86-1; HspcaHeat shock protein HSP 90-alpha; Heat shock 86 kDa; HSP 86; HSP86; Tumor-specific transplantation 86 kDa antigen; TSTA. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8529