Learn More
Biomatik Corporation Mouse Hepatocyte growth factor activator (HGFAC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37044
Encompass_Preferred
Mouse Hepatocyte Growth Factor Activator (Hgfac) Elisa Kit for detecting Hgf Activator in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9R098; Hgfac. Target Synonyms: Hgfac; Hepatocyte growth factor activator; HGF activator; HGFA; EC 3.4.21.-) [Cleaved into: Hepatocyte growth factor activator short chain; Hepatocyte growth factor activator long chain]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8533