Learn More
Biomatik Corporation Mouse hypoxia-inducible factor 1, HIF-1 ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37078
Encompass_Preferred
Mouse Hypoxia-Inducible Factor 1Α, Hif-1Α Elisa Kit for detecting Hypoxia Inducible Factor 1, Alpha Subunit (Basic Helix-Loop-Helix Transcription Factor) in serum, plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40pg/ml. Sensitivity: 0.156pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q61221; Hif1a. Target Synonyms: Hif1aHypoxia-inducible factor 1-alpha; HIF-1-alpha; HIF1-alpha; ARNT-interacting protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8539