Learn More
Biomatik Corporation Mouse ICOS ligand (ICOSLG) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37079
Encompass_Preferred
Mouse Icos Ligand (Icoslg) Elisa Kit for detecting Inducible T-Cell Co-Stimulator Ligand in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9JHJ8; Icoslg. Target Synonyms: Icoslg; B7h2; B7rp1; IcoslICOS ligand; B7 homolog 2; B7-H2; B7-like protein Gl50; B7-related protein 1; B7RP-1; LICOS; CD antigen CD275. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8540