Learn More
Biomatik Corporation Mouse Immunoglobulin A, IgA ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37082
Encompass_Preferred
Mouse Immunoglobulin A, Iga Elisa Kit for detecting Immunoglobulin A, Iga in serum, plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 25-1600ng/ml. Sensitivity: 25ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8541