Learn More
Biomatik Corporation Mouse Immunoglobulin D (IgD) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37083
Encompass_Preferred
Mouse Immunoglobulin D (Igd) Elisa Kit for detecting Immunoglobulin D(Igd) in serum, plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000ng/ml. Sensitivity: 3.9ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8542