Learn More
Biomatik Corporation Mouse Indoleamine 2, 3-dioxygenase 1 (IDO1) ELISA Kit, 96 tests
Catalog No. 501498544
Encompass_Preferred
Mouse Indoleamine 2, 3-Dioxygenase 1 (Ido1) Elisa Kit for detecting Indoleamine 2, 3-Dioxygenase 1 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P28776; Ido1. Target Synonyms: Ido1; Ido; Indo; Indoleamine 2; 3-dioxygenase 1; IDO-1; EC 1.13.11.52; Indoleamine-pyrrole 2; 3-dioxygenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8544 Supplier Biomatik Corporation Supplier No. EKC37090
Mouse Indoleamine 2, 3-Dioxygenase 1 (Ido1) Elisa Kit for detecting Indoleamine 2, 3-Dioxygenase 1 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P28776; Ido1. Target Synonyms: Ido1; Ido; Indo; Indoleamine 2; 3-dioxygenase 1; IDO-1; EC 1.13.11.52; Indoleamine-pyrrole 2; 3-dioxygenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.