Learn More
Biomatik Corporation Mouse inducible nitric oxide synthase, iNOS ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37091
Encompass_Preferred
Mouse Inducible Nitric Oxide Synthase, Inos Elisa Kit for detecting Nitric Oxide Synthase 2, Inducible in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500IU/ml. Sensitivity: 1.95IU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P29477; Nos2. Target Synonyms: Nos2; InoslNitric oxide synthase; inducible; EC 1.14.13.39; Inducible NO synthase; Inducible NOS; iNOS; Macrophage NOS; MAC-NOS; NOS type II; Peptidyl-cysteine S-nitrosylase NOS2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8545