Learn More
Biomatik Corporation Mouse Insulin-like growth factor-binding protein complex acid labile subunit (IGFALS) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37107
Encompass_Preferred
Mouse Insulin-Like Growth Factor-Binding Protein Complex Acid Labile Subunit (Igfals) Elisa Kit for detecting Insulin-Like Growth Factor Binding Protein, Acid Labile Subunit in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P70389; Igfals. Target Synonyms: Igfals; Albs; AlsInsulin-like growth factor-binding protein complex acid labile subunit; ALS. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8550