Learn More
Biomatik Corporation Mouse Interleukin-22 receptor subunit alpha-2 (IL22RA2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37204
Encompass_Preferred
Mouse Interleukin-22 Receptor Subunit Alpha-2 (Il22Ra2) Elisa Kit for detecting Interleukin 22 Receptor, Alpha 2 in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 46.88-3000pg/ml. Sensitivity: 11.7pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q80XF5; Il22ra2. Target Synonyms: Il22ra2Interleukin-22 receptor subunit alpha-2; IL-22 receptor subunit alpha-2; IL-22R-alpha-2; IL-22RA2; Cytokine receptor family type 2; soluble 1; CRF2-S1; Interleukin-22-binding protein; IL-22BP; IL22BP; ZcytoR16. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8561