Learn More
Biomatik Corporation Mouse Keratinocyte Growth Factor (KGF/FGF-7) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37242
Encompass_Preferred
Mouse Keratinocyte Growth Factor (Kgf/Fgf-7) Elisa Kit for detecting Fibroblast Growth Factor 7 (Keratinocyte Growth Factor) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.39pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P36363; Fgf7. Target Synonyms: Fgf7; Fgf-7; Kgf; Fibroblast growth factor 7; FGF-7; Heparin-binding growth factor 7; HBGF-7; Keratinocyte growth factor; KGF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8566