Learn More
Biomatik Corporation Mouse malonyl coenzyme A ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37344
Encompass_Preferred
Mouse Malonyl Coenzyme A Elisa Kit for detecting Malonyl Coenzyme A in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8583