Learn More
Biomatik Corporation Mouse Mast Cell Tryptase (MCT) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37350
Encompass_Preferred
Mouse Mast Cell Tryptase (Mct) Elisa Kit for detecting Mast Cell Tryptase(Mct) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q02844; Tpsab1. Target Synonyms: Tpsab1; Mcpt7; Tryptase; EC 3.4.21.59; Mast cell protease 7; mMCP-7; Tryptase alpha/beta-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8585