Learn More
Biomatik Corporation Mouse myeloperoxidase-antineutrophil cytoplasmic antibody ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37432
Encompass_Preferred
Mouse Myeloperoxidase-Antineutrophil Cytoplasmic Antibody Elisa Kit for detecting Myeloperoxidase-Antineutrophil Cytoplasmic Antibody in serum, plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.156ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8597