Learn More
Biomatik Corporation Mouse NAD (P)H dehydrogenase [quinone] 1 (NQO1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37443
Encompass_Preferred
Mouse Nad (P)H Dehydrogenase [Quinone] 1 (Nqo1) Elisa Kit for detecting Nad(P)H Dehydrogenase, Quinone 1 in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q64669; Nqo1. Target Synonyms: Nqo1; Dia4; Nmo1; Nmor1; NAD (P)H dehydrogenase [quinone] 1; EC 1.6.5.2; Azoreductase; DT-diaphorase; DTD; Menadione reductase; NAD (P)H:quinone oxidoreductase 1; Phylloquinone reductase; Quinone reductase 1; QR1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8601