Learn More
Biomatik Corporation Mouse Neurogranin (NRGN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37473
Encompass_Preferred
Mouse Neurogranin (Nrgn) Elisa Kit for detecting Neurogranin (Protein Kinase C Substrate, Rc3) in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 0.78ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P60761; Nrgn. Target Synonyms: NrgnNeurogranin; Ng; RC3) [Cleaved into: NEUG (55-78)]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8604