Learn More
Biomatik Corporation Mouse Ornithine carbamoyltransferase, mitochondrial (OTC) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37521
Encompass_Preferred
Mouse Ornithine Carbamoyltransferase, Mitochondrial (Otc) Elisa Kit for detecting Ornithine Carbamoyltransferase in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200mU/ml. Sensitivity: 0.78mU/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P11725; Otc. Target Synonyms: Otc; Ornithine carbamoyltransferase; mitochondrial; EC 2.1.3.3; Ornithine transcarbamylase; OTCase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8614