Learn More
Biomatik Corporation Mouse Osteocalcin/Bone gla protein, OT/BGP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37524
Encompass_Preferred
Mouse Osteocalcin/Bone Gla Protein, Ot/Bgp Elisa Kit for detecting Bone Gamma-Carboxyglutamate (Gla) Protein in serum, plasma, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P86546; Bglap. Target Synonyms: Bglap; Osteocalcin; Bone Gla protein; BGP; Gamma-carboxyglutamic acid-containing protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8615