Learn More
Biomatik Corporation Mouse oxytocin, OT ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37537
Encompass_Preferred
Mouse Oxytocin, Ot Elisa Kit for detecting Oxytocin, Ot in serum, plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.6-20pg/ml. Sensitivity: 0.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8618