Learn More
Biomatik Corporation Mouse Perforin/Pore-forming protein, PF/PFP ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKC37556
Encompass_Preferred
Mouse Perforin/Pore-Forming Protein, Pf/Pfp Elisa Kit for detecting Perforin 1 (Pore Forming Protein) in serum, plasma, cell culture supernates. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P10820; Prf1. Target Synonyms: Prf1; Pfp; Perforin-1; P1; Cytolysin; Lymphocyte pore-forming protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-149-8624